MyoCult™ Differentiation Kit (Human)
Proveedor: STEMCELL Technologies
05965
STMC05965EA 314 EUR
STMC05965
MyoCult™ Differentiation Kit (Human)
Medios de cultivo celular
Medium for the differentiation of human skeletal muscle progenitor cells (myoblasts) into myotubes.
MyoCult™ Differentiation Kit (Human) includes MyoCult™ Differentiation Medium and Animal Component-Free (ACF) Cell Attachment Substrate. MyoCult™ Differentiation Medium is a serum-based medium specifically formulated for the in vitro differentiation of human skeletal muscle progenitor cells (myoblasts) into myotubes. MyoCult™ Differentiation Medium is designed for use with the ACF Cell Attachment Substrate, which has been specifically tested for its ability to support optimal adherence and myotube morphology. This medium is suitable for the differentiation of myogenic progenitor cells previously expanded in serum-containing or serum-free media (e.g. using the MyoCult™-SF Expansion Kit). CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate. The Product may be used for research purposes only. Please contact STEMCELL for any clinical or commercial application of the Product.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').