Anti-Adherence Rinsing Solution

Proveedor: STEMCELL Technologies
07010
STMC07010EA 56 EUR
STMC07010
Anti-Adherence Rinsing Solution
Medios para separación celular
Anti-Adherence Rinsing Solution is a surfactant solution for pre-treating cultureware to reduce surface tension and prevent cell adhesion. It can be used with plates, flasks, pipettes, strainers and other cultureware.

Adherence rinsing solution is required for use with the following:
- AggreWell™ plates (e.g. Catalog MSPP-34811)
- Organoid growth media, including HepatiCult™ (Catalog MSPP-06030)
- PancreaCult™ (Catalog MSPP-06040)

Application: Cancer, neuroscience, stem cell biology
Species: Human, mouse, non human primate, other, rat

Precaución: Note: formerly known as AggreWell™More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR