STEMdiff™ Cerebral Organoid Kit
Proveedor: STEMCELL Technologies
08570
STMC08570EA 410 EUR
STMC08570
STEMdiff™ Cerebral Organoid Kit
Medios de cultivo celular
Culture medium kit for establishment and maturation of human cerebral organoids.
Generate self-organized, pluripotent stem cell (PSC)-derived neural organoids with a cellular composition and structural organization representative of the developing human brain. These defined, serum-free cell culture media and the simple, four-stage protocol are based on the formulation published by Lancaster et al. to more reliably generate cerebral organoids.
Beginning with an embryoid body (EB) formation step followed by expansion of neuroepithelia, organoids generated using STEMdiff™ Cerebral Organoid Kit feature cortical-like regions, including the ventricular zone (PAX6+/SOX2+/Ki-67+), outer subventricular zone (Ki-67+/p-Vimentin+), intermediate zone (TBR2+), and cortical plate (CTIP2+/MAP2+/TBR1+), which layer in similar orientations as those observed in vivo. For extended culture periods (> 40 days), the components required for maturation can be purchased as STEMdiff™ Cerebral Organoid Maturation Kit.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').