Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (PE)
Proveedor: STEMCELL Technologies
100-0311 100-0312 100-0313
STMC100-0311EA 0 EUR
STMC100-0311 STMC100-0312 STMC100-0313
Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (PE)
Anticuerpos
Mouse monoclonal IgG2a, antibody against human, rhesus, cynomolgus HLA-DR, PE-conjugated.
  • B cells
  • Macrophages
  • Monocytes
  • T cells
The L243 antibody reacts with human HLA-DR, the major histocompatibility complex (MHC) class II receptor and a member of the immunoglobulin protein superfamily. HLA-DR is a heterodimeric transmembrane glycoprotein comprising a 36-kDa α subunit associated non-covalently with a 27-kDa β subunit and is expressed on the surface of antigen-presenting cells such as B cells, activated T cells, monocytes, macrophages and dendriticMore Product Information