HepatiCult™ Organoid Growth Medium (Human)
Proveedor: STEMCELL Technologies
100-0385
STMC100-0385EA 403 EUR
STMC100-0385
HepatiCult™ Organoid Growth Medium (Human)
Medios de cultivo celular
Culture medium for long-term expansion and maintenance of human liver organoids.
Reliably expand, and maintain human liver organoids long-term as a model system for studying liver biology, disease modeling, drug discovery, and therapeutic screening. Use HepatiCult™ Organoid Growth Medium (OGM) (Human) to establish hepatic organoids from human liver tissue, or PSC-derived hepatic cells generated with the STEMdiff™ Hepatocyte Kit. Available either individually or as part of HepatiCult™ Organoid Kit (Human), HepatiCult™ OGM can be used to culture and expand liver organoids for future experimentation or bio-banking.
Organoids initiated in HepatiCult™ Organoid Initiation Medium (Human) and expanded in HepatiCult™ OGM express self-renewal (LGR5, AXIN2), ductal (KRT19, SOX9, KRT7), and hepatic progenitor (HNF4A) markers. These organoids can also be differentiated using HepatiCult™ Organoid Differentiation Medium (Human) to mature organoids that demonstrate hepatic functionality, including CYP3A4 activity. Use HepatiCult™ OGM to culture organoids in a range of formats, including in Corning® Matrigel® domes for routine culture, in dilute Matrigel® suspensions in spinner flasks for scale-up, in 96-well plates for high-throughput screening applications, and as organoid-derived two-dimensional monolayers on Transwell® inserts for disease-modeling applications.
The organoids can also be cryopreserved for later use, remaining highly viable and retaining their capacity for further differentiation when thawed and cultured in HepatiCult™ OGM.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').