ImmunoCult™ Human Treg Differentiation Supplement
Proveedor: STEMCELL Technologies
10977
STMC10977EA 289 EUR
STMC10977
ImmunoCult™ Human Treg Differentiation Supplement
Medios de cultivo celular
Culture supplement for the differentiation of human naïve CD4+ T cells into tregs.
ImmunoCult™ Human Treg Differentiation Supplement contains a human cytokine and small molecule formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into regulatory T cells (Tregs). ImmunoCult™ Human Treg Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium and ImmunoCult™ Human CD3/CD28 T Cell Activator.
This product is recommended for:
Research into the regulation of human naïve T cell differentiation and Treg function
Development of procedures to expand and polarise naïve T cells into Tregs
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').