RosetteSep™ HLA T Cell Enrichment Cocktails
Proveedor: STEMCELL Technologies
15061HLA 15081HLA
STMC15061HLAEA 1193 EUR
STMC15061HLA STMC15081HLA
RosetteSep™ HLA T Cell Enrichment Cocktails
Medios para separación celular
Immunodensity isolation of untouched T cells for HLA analysis.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Isolated cells are untouched
The RosetteSep™ HLA T Cell Enrichment Cocktail is designed to isolate T cells from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes (TAC) recognising non-T cells and red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (STMC15705) or Lymphoprep™ (STMC07801), the unwanted cells pellet along withMore Product Information