RosetteSep™ Human Multiple Myeloma Cell Enrichment Cocktails
Proveedor: STEMCELL Technologies
15169 15129
STMC15169EA 2591 EUR
STMC15169 STMC15129
RosetteSep™ Human Multiple Myeloma Cell Enrichment Cocktails
Medios para separación celular
Immunodensity isolation of untouched plasma cells for multiple myeloma research.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
The RosetteSep™ Human Multiple Myeloma Cell Enrichment Cocktail is designed to enrich multiple myeloma cells (B cell and plasma cells) from fresh bone marrow aspirates by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD2, CD14, CD33, CD41, CD45RA, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over aMore Product Information