EasySep™ Human PE Positive Selection Kit II
Proveedor: STEMCELL Technologies
17664 17694 17664RF
STMC17664EA 821 EUR
STMC17664 STMC17694 STMC17664RF
EasySep™ Human PE Positive Selection Kit II
Medios para separación celular
Immunomagnetic positive selection of human cells labeled with PE-conjugated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human PE Positive Selection Kit II is designed to isolate cells that are labeled with PE (phycoerythrin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognising PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.