EasySep™ APC Positive Selection Kit II
Proveedor: STEMCELL Technologies
17681 17681RF
STMC17681EA 679 EUR
STMC17681 STMC17681RF
EasySep™ APC Positive Selection Kit II
Medios para separación celular
Immunomagnetic positive selection of any cells labeled with APC-conjugated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ APC Positive Selection Kit II is designed to isolate cells that are labeled with APC (allophycocyanin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognising APC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.