EasySep™ Human CD271 Positive Selection Kit II
Proveedor: STEMCELL Technologies
17849
STMC17849EA 849 EUR
STMC17849
EasySep™ Human CD271 Positive Selection Kit II
Medios para separación celular
Immunomagnetic positive selection of human CD271+ cells.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human CD271 Positive Selection Kit II is designed to isolate CD271+ cells from fresh human bone marrow by positive selection. Desired cells are targeted with antibody complexes recognising CD271 and dextran-coated magnetic particles. The kit also contains an antibody to human Fc receptor to minimize nonspecific binding. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube whileMore Product Information