EasySep™ HLA Chimerism Whole Blood Myeloid Positive Selection Kits
Proveedor: STEMCELL Technologies
17884 17884RF
STMC17884EA 863 EUR
STMC17884 STMC17884RF
EasySep™ HLA Chimerism Whole Blood Myeloid Positive Selection Kits
Medios para separación celular
Column-free immunomagnetic positive selection of human CD33+ and CD66b+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
Isolate highly purified CD33+ and CD66b+ myeloid cells, including myeloid progenitor cells, granulocytes, and monocytes, from fresh whole blood prior to chimerism testing.
Desired cells are targeted with tetrameric antibody complexes recognising CD33, CD66b, and dextran-coated magnetic particles, and separated without the use of columns through immunomagnetic positive selection using an EasySep™More Product Information