EasySep™ Human Neutrophil Isolation Kits
Proveedor: STEMCELL Technologies
17957 17957RF
STMC17957EA 1377 EUR
STMC17957 STMC17957RF
EasySep™ Human Neutrophil Isolation Kits
Medios para separación celular
Immunomagnetic isolation of untouched human neutrophils.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human Neutrophil Isolation Kit is designed to isolate neutrophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-neutrophils and magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. TheMore Product Information