EasySep™ 'The Big Easy' Magnet
Proveedor: STEMCELL Technologies
18001
STMC18001EA 1800 EUR
STMC18001
EasySep™ 'The Big Easy' Magnet
Imánes
The EasySep™ 'The Big Easy' magnet is designed for cell separation procedures using EasySep™ reagents when processing a large number of cells, or when using RoboSep™.
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 14 ml (17×95 mm) polystyrene tube
'The Big Easy' magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ magnetic particles, without the use of columns.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of themMore Product Information