EasySep™ Easy 50 Magnet
Proveedor: STEMCELL Technologies
18002
STMC18002EA 1777 EUR
STMC18002
EasySep™ Easy 50 Magnet
Imánes
The EasySep™ Easy 50 magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a large number of cells (up to 4×10⁹ cells).
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 50 ml conical tube
The EasySep™ Easy 50 magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with magnetic EasySep™ particles, without the use of columns.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way,More Product Information