EasySep™ Human Plasmacytoid DC Enrichment Kits
Proveedor: STEMCELL Technologies
19062 19062RF
STMC19062EA 1356 EUR
STMC19062 STMC19062RF
EasySep™ Human Plasmacytoid DC Enrichment Kits
Medios para separación celular
Immunomagnetic isolation of untouched human plasmacytoid dendritic cells (pDC)
  • Fast, easy-to-use and column-free
  • Up to 97% purity
  • Untouched, viable cells
The EasySep™ Human Plasmacytoid DC Enrichment Kit is designed to isolate plasmacytoid dendritic cells (pDC) from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-pDCs and dextran-coated magnetic particles. The labelled cells are separated using the EasySep™ magnet without the use of columns.More Product Information