Hausser Scientific™ Hemocytometer Cover Glasses
Proveedor: STEMCELL Technologies
200-0641
STMC200-0641EA 182 EUR
STMC200-0641
Hausser Scientific™ Hemocytometer Cover Glasses
Cubreobjetos
Count cells with hemocytometer cover glasses designed specifically for use with the Hausser Scientific™ Bright-Line Hemocytometer.
  • Replacement cover glasses
These cover glasses facilitate manual cell counting at a fraction of the cost and footprint of automated cell counters.
Envase: They are available in packs of 10.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove theMore Product Information