Anti-CD11c Hamster (Armenian) Monoclonal Antibody [clone: N418] (Biotinilated)
Proveedor: STEMCELL Technologies
60002BT 100-0442 100-0441
STMC60002BTEA 393 EUR
STMC60002BT STMC100-0442 STMC100-0441
Anti-CD11c Hamster (Armenian) Monoclonal Antibody [clone: N418] (Biotinilated)
Anticuerpos
Hamster (Armenian) monoclonal IgG antibody against mouse CD11c, biotin-conjugated.
  • Dendritic cells
The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor. Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cellMore Product Information