Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP-Cy5.5)

Proveedor: STEMCELL Technologies
60005PS 60005PS.1
STMC60005PSEA 0 EUR
STMC60005PS STMC60005PS.1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP-Cy5.5)
Anticuerpos
Mouse monoclonal IgG1 antibody against human, chimpanzee CD19, PerCP-Cy5.5-conjugated.

  • B cells

The HIB19 antibody reacts with CD19, an ~95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells. Expression is down-regulated but persists in terminally differentiated plasma cells. CD19 is also found on follicular dendritic cells. By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation andMore Product Information
Order Now

ESPECIFICACIONES

Nombre del antígeno CD19
Environmentally Preferable
Clonalidad Monoclonal
Conjugado PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Clon HIB19
Reactividad Chimpanzee,Human
Huésped Mouse
ID del gen 930
Isotipo IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR