Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (APC)
Proveedor: STEMCELL Technologies
60018AZ 60018AZ.1 100-0467
STMC60018AZEA 328 EUR
STMC60018AZ STMC60018AZ.1 STMC100-0467
Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (APC)
Anticuerpos
Mouse monoclonal IgG1 antibody against human, chimpanzee CD45, APC-conjugated.
  • Granulocytes
  • Subsets
  • Hematopoietic stem
  • Progenitor cells
  • Leukocytes
  • Lymphocytes
  • Myeloid cells
The HI30 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell types. CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes,More Product Information