Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631] (Alexa Fluor® 488)
Proveedor: STEMCELL Technologies
60061AD 60061AD.1
STMC60061ADEA 513 EUR
STMC60061AD STMC60061AD.1
Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631] (Alexa Fluor® 488)
Anticuerpos
Rat monoclonal IgM antibody against human, mouse, rat SSEA-3, Alexa Fluor® 488-conjugated.
  • Pluripotent stem cells
The MC-631 antibody reacts with a galactosylgloboside epitope on the stage-specific embryonic antigen-3 (SSEA-3), which is expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey ES cell lines. No immunoreactivity is evident with undifferentiated mouse EC, EG and ES cells. Expression of SSEA-3 is down regulatedMore Product Information