Anti-CD68 Mouse Monoclonal Antibody [clone: Y1/82A] (Alexa Fluor® 488)

Proveedor: STEMCELL Technologies
60105AD 60105AD.1
STMC60105ADEA 404 EUR
STMC60105AD STMC60105AD.1
Anti-CD68 Mouse Monoclonal Antibody [clone: Y1/82A] (Alexa Fluor® 488)
Anticuerpos
Mouse monoclonal IgG2b antibody against human CD68, Alexa Fluor® 488-conjugated.

  • Granulocytes
  • Subsets
  • Myeloid cells

The Y1/82A antibody reacts with human CD68, an ~110 kDa type 1 transmembrane glycoprotein and member of the sialomucin, LAMP and scavenger receptor families. The heavily glycosylated extracellular domain of CD68, which contains a praline-rich spacer region separating mucin and LAMP domains, binds low-density lipoprotein and certain lectins and selectins, though the function of CD68 remains unclear. CD68 is highly expressed in lysosomes,More Product Information
Order Now

ESPECIFICACIONES

Nombre del antígeno CD68
Environmentally Preferable
Clonalidad Monoclonal
Conjugado Alexa Fluor® 488
Clon Y1/82A
Reactividad Human
Huésped Mouse
ID del gen 968
Isotipo IgG2b kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR