Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (APC)

Proveedor: STEMCELL Technologies
60109AZ 60109AZ.1
STMC60109AZEA 0 EUR
STMC60109AZ STMC60109AZ.1
Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (APC)
Anticuerpos
Rat monoclonal IgG2a antibody against mouse CD62L (L-selectin), APC-conjugated.

  • T cells

The MEL-14 antibody reacts with murine CD62L (L-Selectin or LECAM-1), an ~74 - 95 kDa type I glycoprotein expressed on the surface of a majority of leukocytes. CD62L mediates leukocyte-endothelial cell interactions through its association with ligands such as CD34, glyCAM-1, MAdCAM-1 and PSGL-1. It functions to facilitate lymphocyte rolling on the vascular endothelium, trafficking of lymphocytes to the lymph nodes, and homing of lymphocytes and neutrophils to sitesMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CD62L
Nombre del antígeno L-Selectin
Environmentally Preferable
Clonalidad Monoclonal
Conjugado APC (Allophycocyanin)
Clon MEL-14
Reactividad Mouse
Huésped Rat
ID del gen 20343
Isotipo IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR