Human/Mouse Recombinant FGF-8B, ACF
Proveedor: STEMCELL Technologies
78204 78204.1 78204.2
STMC78204EA 208 EUR
STMC78204 STMC78204.1 STMC78204.2
Human/Mouse Recombinant FGF-8B, ACF
Proteínas y Péptidos
Fibroblast growth factor 8B (FGF-8B) is a member of the fibroblast growth factor (FGF) family and is an isoform of FGF-8. Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman and Klagsbrun; Kimelman and Kirschner) and are involved in a variety of biological processes, including cell proliferation, differentiation, survival, and apoptosis (Folkman and Klagsbrun; Klagsbrun; Rifkin and Moscatelli). FGF-8B signals through FGF receptors (FGFRs) to activate PI3K and MAPK pathways. FGF-8B regulates gastrulation, epithelial-mesenchymal transition, and mesenchymal to epithelial differentiation during embryonic development. FGF-8B has also been found in peripheral blood leukocytes and healthy bone marrow samples (Mattila and Härkönen). FGF-8B has mitogenic effects on somatic cells in the germinal epithelium and is expressed in adult mouse ovarian cells and tissues, which suggests that it regulates maturation of oocytes and seminiferous epithelium in testes (Valve et al.). This product is animal component-free.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
ESPECIFICACIONES
Nombre de proteína / péptido FGF-8B, ACF
Environmentally Preferable
Tipo de proteína / péptido Recombinant
Especies Human,Mouse
Fuente
Pureza ≥95%