Atlas® NBE SPE Columns
Proveedor: Tecan
30174231 30174239
TCAT30174231EA 0 EUR
TCAT30174231 TCAT30174239
Atlas® NBE SPE Columns
Columnas de cromatografía
A Hydrophilic/Lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species along with a unique NBE column platform. A direct alternative to Waters Oasis® HLB µElution.
  • Mode: Reversed-phased
Atlas® a hydrophilic-lipophilic balanced polymeric sorbent is a N-vinylpyrrolidone-divinylbenzene co-polymer. It provides selectivity for both polar and non-polar analytes thus allowing the user to extract a variety of samples with ease. The divinylbenzene lipophilic back bone providesMore Product Information