SPE Column, NBE OFXQ
Proveedor: Tecan
635-0031R-NBE 635-0051R-NBE
TCAT635-0031R-NBEEA 0 EUR
TCAT635-0031R-NBE TCAT635-0051R-NBE
SPE Column, NBE OFXQ
Columnas de cromatografía
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
  • Ion-exchange
  • Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.