Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Proveedor: Tecan
94000001 94100001 94200001 94300001 94400001
TCAT94000001EA 0 EUR
TCAT94000001 TCAT94100001 TCAT94200001 TCAT94300001 TCAT94400001
Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Lectores de microplacas
Spark Cyto is the first multimode reader with live cell imaging and real time cytometry to offer as much information as possible from the same cells, under the same conditions.
  • Live cell imaging and real-time cytometry for 2D and 3D cell models
  • Temperature and gas control
  • Lid Lifter™
  • Enhanced Absorbance, Fluorescence and Luminescence modules
  • Fusion optics - combine the flexibility of monochromators with the sensitivity of filters
  • ALPHA technology
  • Fluorescence filter top/bottom
  • DNA-, RNA- & protein quantification
  • Low sample volume: 2 µl
  • Up to 16 sample