N(α)-Acetyl-L-tryptophan ≥98.0% (by HPLC)

Proveedor: TCI

A0121-1G A0121-25G
TCIAA0121-1GEA 17 EUR
TCIAA0121-1G TCIAA0121-25G
N(α)-Acetyl-L-tryptophan ≥98.0% (by HPLC)
N(α)-Acetyl-L-tryptophan
Formula: C₁₃H₁₄N₂O₃
Número MDL: MFCD00065976
Núm. CAS: 1218-34-4
EINECS: 214-935-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2c0259ddf6a16c34c8251913c0e67db49175ef2&vid=e2c0259ddf6a16c34c8251913c0e67db49175ef2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D +25.0 to +27.0 deg (C=1, 1mol/L NaOH sol.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR