L-Adrenaline bitartrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

A0175-1G A0175-25G A0175-5G
TCIAA0175-1GEA 34 EUR
TCIAA0175-1G TCIAA0175-25G TCIAA0175-5G
L-Adrenaline bitartrate ≥98.0% (by HPLC, titration analysis)
L-Adrenaline bitartrate
Formula: C₉H₁₃NO₃
Temperatura de almacenaje: Nevera
Número MDL: MFCD00035077
Núm. CAS: 51-42-3
EINECS: 200-097-1
UN: 2811
ADR: 6.1,II

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) 98.0 to 102.0 %
Specific rotation [a]20/D -15.0 to -18.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR