DL-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 3-Aminobutanoic acid

A0281-1G A0281-25G A0281-5G
TCIAA0281-1GEA 16 EUR
TCIAA0281-1G TCIAA0281-25G TCIAA0281-5G
DL-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)
DL-3-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.1198 g/mol
Número MDL: MFCD00008087
Núm. CAS: 541-48-0
EINECS: 208-783-2
Índice Merck: 14,00429
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0555aae9bc9c217195be49d61722032ae1aee927&vid=0555aae9bc9c217195be49d61722032ae1aee927&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR