DL-3-Aminoisobutyric acid hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 3-Amino-2-methylpropanoic acid, BAIBA

A0324-1G A0324-5G
TCIAA0324-1GEA 45 EUR
TCIAA0324-1G TCIAA0324-5G
DL-3-Aminoisobutyric acid hydrate ≥98.0% (by titrimetric analysis)
DL-3-Aminoisobutyric acid hydrate
Formula: C₄H₉NO₂·H₂O
MW: 103.12 g/mol
Número MDL: MFCD00008145
Núm. CAS: 144-90-1
EINECS: 205-644-8

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0% (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 15.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR