Potassium amylxanthate ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Amylxanthic Acid Potassium Salt, Potassium O-Amyl Dithiocarbonate

A0461-25G
TCIAA0461-25GEA 105 EUR
TCIAA0461-25G
Potassium amylxanthate ≥97.0% (by titrimetric analysis)
Potassium amylxanthate
Formula: C₆H₁₂OS₂
Número MDL: MFCD00059150
Núm. CAS: 2720-73-2
EINECS: 220-329-5

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Solubility in Water very faint turbidity
Sensitiveness Colorimetric Test : Turn Pale with 5ppm Molybdenum


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR