Sodium anthraquinone-2-sulphonate monohydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

A0508-25G A0508-500G
TCIAA0508-25GEA 17 EUR
TCIAA0508-25G TCIAA0508-500G
Sodium anthraquinone-2-sulphonate monohydrate ≥98.0% (by HPLC, titration analysis)
Sodium anthraquinone-2-sulphonate monohydrate
Formula: C₁₄H₈O₅S
Número MDL: MFCD00149068
Núm. CAS: 153277-35-1
EINECS: 205-009-5

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR