2-Ácido arsanilico ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: o-Arsanilic acid, 2-Aminophenylarsonic acid, 2-Aminobenzenearsonic acid

A0529-25G A0529-5G
TCIAA0529-25GEA 126 EUR
TCIAA0529-25G TCIAA0529-5G
2-Ácido arsanilico ≥98.0% (by titrimetric analysis)
2-Ácido arsanilico
Formula: C₆H₈AsNO₃
Número MDL: MFCD00007630
Núm. CAS: 2045-00-3
EINECS: 218-064-5
UN: 3465
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31a929f6483ea15a22ed7473cd4c4c9b657831b2&vid=31a929f6483ea15a22ed7473cd4c4c9b657831b2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 155.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR