Atropine sulphate monohydrate ≥99.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

A0550-25G A0550-5G
TCIAA0550-25GEA 88 EUR
TCIAA0550-25G TCIAA0550-5G
Atropine sulphate monohydrate ≥99.0% (by titrimetric analysis)
Atropine sulfate monohydrate
Formula: C₁₇H₂₃NO₃
Pto. de fusión: 194°C
Número MDL: MFCD00074815
Núm. CAS: 5908-99-6
EINECS: 200-235-0
UN: 1544
ADR: 6.1,II

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR