2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

A0896-25G A0896-500G
TCIAA0896-25GEA 16 EUR
TCIAA0896-25G TCIAA0896-500G
2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt ≥98.0% (by HPLC, total nitrogen)
2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt
Formula: C₇H₇ClNNaO₃S
Número MDL: MFCD00070551
Núm. CAS: 6627-59-4
EINECS: 229-596-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6820ec1bbc8a080b42d6b70a54ec846e44fc4005&vid=6820ec1bbc8a080b42d6b70a54ec846e44fc4005&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Water almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR