Dimethyl-1,8-anthracenedicarboxylate ≥95.0% (por GC)

Proveedor: TCI

Sinónimos: 1,8-Anthracenedicarboxylic Acid Dimethyl Ester

A1433-5G A1433-1G
TCIAA1433-5GEA 783 EUR
TCIAA1433-5G TCIAA1433-1G
Dimethyl-1,8-anthracenedicarboxylate ≥95.0% (por GC)
Dimethyl-1,8-anthracenedicarboxylate
Formula: C₁₈H₁₄O₄
Número MDL: MFCD01321126
Núm. CAS: 93655-34-6

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 102.0 to 106.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR