3-Amino-4-pyrazolecarbonitrile ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A1594-25G A1594-5G
TCIAA1594-25GEA 82 EUR
TCIAA1594-25G TCIAA1594-5G
3-Amino-4-pyrazolecarbonitrile ≥98.0% (by GC, titration analysis)
3-Amino-4-pyrazolecarbonitrile
Formula: C₄H₄N₄
Número MDL: MFCD00005237
Núm. CAS: 16617-46-2
EINECS: 240-665-6

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 174.0 to 177.0 deg-C
Solubility in hot Methanol very faint turbidity

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR