(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A1623-1G A1623-5G
TCIAA1623-1GEA 16 EUR
TCIAA1623-1G TCIAA1623-5G
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol
Formula: C₉H₁₁NO
Número MDL: MFCD00216656
Núm. CAS: 136030-00-7

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR