N-(4-Aminobenzoyl)-β-alanine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 3-(4-Aminobenzamido)propionic Acid, N-(4-Aminobenzoyl)-beta-alanine

A2347-25G A2347-5G
TCIAA2347-25GEA 28 EUR
TCIAA2347-25G TCIAA2347-5G
N-(4-Aminobenzoyl)-β-alanine ≥98.0% (by HPLC, titration analysis)
N-(4-Aminobenzoyl)-beta-alanine
Formula: C₁₀H₁₂N₂O₃
MW: 208.22 g/mol
Número MDL: MFCD00009805
Núm. CAS: 7377-08-4

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR