9-Anthrylmethyl methacrylate ≥98.0% (by HPLC)

Proveedor: TCI
Atención

A2645-1G
TCIAA2645-1GEA 112 EUR
TCIAA2645-1G
9-Anthrylmethyl methacrylate ≥98.0% (by HPLC)
9-Anthrylmethyl methacrylate
Formula: C₁₉H₁₆O₂
Número MDL: MFCD00673247
Núm. CAS: 31645-35-9

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR