6-Amino-1-methyl-5-(propylamino)uracil ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

A2715-1G
TCIAA2715-1GEA 105 EUR
TCIAA2715-1G
6-Amino-1-methyl-5-(propylamino)uracil ≥98.0% (by HPLC, titration analysis)
6-Amino-1-methyl-5-(propylamino)uracil
Formula: C₈H₁₄N₄O₂
Número MDL: MFCD01074825
Núm. CAS: 125092-42-4

Order Now

Resultados test especificaciones

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR