3-Amino-N-(tert-butoxycarbonyl)benzylamine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A2718-1G A2718-5G
TCIAA2718-1GEA 160 EUR
TCIAA2718-1G TCIAA2718-5G
3-Amino-N-(tert-butoxycarbonyl)benzylamine ≥98.0% (by GC, titration analysis)
3-Amino-N-(tert-butoxycarbonyl)benzylamine
Formula: C₁₂H₁₈N₂O₂
Número MDL: MFCD04974010
Núm. CAS: 147291-66-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 48.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR