2-Amino-4'-chlorobiphenyl ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A3034-1G A3034-5G
TCIAA3034-1GEA 60 EUR
TCIAA3034-1G TCIAA3034-5G
2-Amino-4'-chlorobiphenyl ≥98.0% (by GC, titration analysis)
2-Amino-4'-chlorobiphenyl
Formula: C₁₂H₁₀ClN
Número MDL: MFCD03424659
Núm. CAS: 1204-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb0cd51940068f81f9c76cbb7e7220f5ae1e0e04&vid=fb0cd51940068f81f9c76cbb7e7220f5ae1e0e04&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR