2-[2-(2-Aminoethoxy)ethoxy]acetic acid ≥96.0% (by titrimetric analysis)

Proveedor: TCI

A3120-250MG A3120-1G
TCIAA3120-250MGEA 60 EUR
TCIAA3120-250MG TCIAA3120-1G
2-[2-(2-Aminoethoxy)ethoxy]acetic acid ≥96.0% (by titrimetric analysis)
2-[2-(2-Aminoethoxy)ethoxy]acetic acid
Formula: C₆H₁₃NO₄
Temperatura de almacenaje: Congelador
Número MDL: MFCD13185897
Núm. CAS: 134978-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=634c19e3b472f09c1c7deef0737e13d8d1b30992&vid=634c19e3b472f09c1c7deef0737e13d8d1b30992&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR