Disodium-1,4-butanedisulphonate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 1,4-Butanedisulphonic Acid Disodium Salt, Sodium butane-1,4-disulphonate

B0676-10G
TCIAB0676-10GEA 208 EUR
TCIAB0676-10G
Disodium-1,4-butanedisulphonate ≥98.0% (by titrimetric analysis)
Disodium-1,4-butanedisulphonate
Formula: C₄H₁₀O₆S₂
Número MDL: MFCD00059839
Núm. CAS: 36589-61-4

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 % (calcd.on dried substance)
Solubility in Water almost transparency
Drying loss max. 2.0 %
Sodium Sulfate max. 1.0 % (calcd.on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR