3,3'-Dichloro-N,N'-bis(4-methoxybenzylidene)benzidine ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

B0878-25G
TCIAB0878-25GEA 182 EUR
TCIAB0878-25G
3,3'-Dichloro-N,N'-bis(4-methoxybenzylidene)benzidine ≥98.0% (by titrimetric analysis)
3,3'-Dichloro-N,N'-bis(4-methoxybenzylidene)benzidine
Formula: C₂₈H₂₂Cl₂N₂O₂
Número MDL: MFCD00059294
Núm. CAS: 3540-78-1

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Chloroform almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR