2,2'-[(1,1-Dimetiletil)imino]bisetanol ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

B1194-25ML B1194-500ML
TCIAB1194-25MLEA 16 EUR
TCIAB1194-25ML TCIAB1194-500ML
2,2'-[(1,1-Dimetiletil)imino]bisetanol ≥97.0% (by titrimetric analysis)
2,2'-[(1,1-Dimetiletil)imino]bisetanol
Formula: C₈H₁₉NO₂
Pto. Ebullición: 161°C
Número MDL: MFCD00020584
Núm. CAS: 2160-93-2
EINECS: 218-480-7
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fe53fb337352e422b3d10ed4f952f66b0aef453&vid=6fe53fb337352e422b3d10ed4f952f66b0aef453&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to lump
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR