Bis(methylenedithio)tetrathiafulvalene for organic electronics

Proveedor: TCI

Sinónimos: BMDT-TTF

B1218-100MG
TCIAB1218-100MGEA 228 EUR
TCIAB1218-100MG
Bis(methylenedithio)tetrathiafulvalene for organic electronics
Bis(methylenedithio)tetrathiafulvalene
Formula: C₈H₄S₈
Número MDL: MFCD00059697
Núm. CAS: 68550-20-9

Order Now

Resultados test especificaciones

Appearance Red to Dark red to Brown powder to crystal
Elemental analysis (Sulfuride) 69.0 to 73.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR