9-Bromo-9-phenylfluorene ≥96.0% (por GC)

Proveedor: TCI
Peligro

B1702-25G B1702-5G
TCIAB1702-25GEA 270 EUR
TCIAB1702-25G TCIAB1702-5G
9-Bromo-9-phenylfluorene ≥96.0% (por GC)
9-Bromo-9-phenylfluorene
Formula: C₁₉H₁₃Br
Número MDL: MFCD00075522
Núm. CAS: 55135-66-5
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62389385f6a401fc7bec8178413d1396c58f3704&vid=62389385f6a401fc7bec8178413d1396c58f3704&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 98.0 to 102.0 deg-C
Solubility in Acetone almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR